Learn More
Bullard™ PA30 Series Complete PAPR Assemblies
SDP
Catalog No. 19167755 Shop All Bullard Company Products
Change view
Click to view available options
Loose-fitting facepiece system with standard belt; Not intrinsically safe
Catalog No.
19-167-755 Loose-fitting facepiece system with standard belt; Not intrinsically safe
1 options
This item is not returnable. View return policy
Catalog No. 19-167-755 Supplier Bullard™ Supplier No. PA30LFLSB
This item is not returnable. View return policy
Choose from decon-friendly hospital preparedness design, or any one of numerous intrinsically safe models
Sold as complete assemblies. For descriptions of individual components, see separate entries.
Hospital Preparedness System
  • Comes with Tychem™ SL hood, protective breathing tube cover, vinyl decon belt, and one set of NBC canisters Smooth, “decon-friendly” housing is sealed and gasketed, with downward slopes for easy runoff
  • Rechargeable NiMH battery packs are engineered inside the profile of the housing to provide even weight distribution and eliminate battery cables that may entangle or short One battery lasts up to five hours 77dBA audible low-voltage alarm inside breathing tube connector sends signal to the headphones
  • Airflow: >8cfm
Intrinsically Safe Models
  • All models feature PA3IS intrinsically safe blower with battery, breathing tube, quick charger, low-voltage alarm, and airflow indicator Unit with solvent-resistant lens suits painting applications
  • RT1 and RT2 hoods with extra-large lens provide high-performance results using an inflatable neck cuff design that provides positive custom seal around user's neck For more information about individual hood features and benefits, see separate descriptions Battery provides 6-7 hours of run time
  • Airflow: >7cfm
Specifications
Battery Life 8 to 10 hours
Product Type PAPR System with Hood
For Use With (Application) HEPA, Organic Vapor, Acid Gas Protection
Includes Two batteries, PA3 blower, breathing tube, one loose-fitting facepiece, quick charger, low voltage alarm and air flow indicator, Standard Comfort Belt
Model PA30LFLSB
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.