Learn More
Bullard™ Replacement Breathing Tube
SDP
Catalog No. 189994435 Shop All Bullard Company Products
Change view
Click to view available options
For Use With (Equipment):
Bullard breathing tube assemblies
Catalog No. For Use With (Equipment)
18-999-4435 Bullard breathing tube assemblies
1 options
Catalog No. 18-999-4435 Supplier Bullard™ Supplier No. 88VXBT
Add to Cart
Add to Cart
Breathing tube for use with 88VX respirator.
  • Replacement part
  • For use with 88VX respirator
Specifications
Product Type Breathing Tube
For Use With (Equipment) Bullard breathing tube assemblies
Includes Threaded hose connectors
Model 88VXBT
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.