Learn More
Bullard™ Breathing Tube Assembly for PA40 Powered Air-Purifying Respirators
SDP
Catalog No. 19321650 Shop All Bullard Company Products
This item is not returnable. View return policy
Catalog No. 19-321-650 Supplier Bullard™ Supplier No. PA4BT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement parts for use with PA40 powered air-purifying respirator systems.
  • Component/replacement part for PA40 PAPR system
  • Fits full facepiece respirator
  • Standard length measures 33 in.
Specifications
Product Type Breathing Tube
For Use With (Equipment) PA40 Series PAPR System
Includes Swivel elbow connection
Model PA4BT
Length (Metric) 83.8 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.