Learn More
Bullard™ RT Series Tychem™ Hood Respirator Assemblies
SDP
Catalog No. 19161222 Shop All Bullard Company Products
Change view
Click to view available options
Includes:
RT1 hood, 20BT breathing tube, F30 flow control
RT2 hood, 20BT breathing tube, F30 flow control
RT3 hood, 20BT breathing tube, F30 flow control
RT3 hood, RTBT breathing tube
RT3 hood, RTBT breathing tube, waist strap
RT4 hood, 20BT breathing tube, F30 flow control
RT4 hood, RTBT breathing tube
RT4 hood, RTBT breathing tube, waist strap
Material:
Tychem QC
Tychem SL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Material
19-161-222 RT3 hood, RTBT breathing tube Tychem QC
19-161-223 RT4 hood, RTBT breathing tube Tychem SL
19-321-720 RT3 hood, RTBT breathing tube, waist strap Tychem QC
19-321-667 RT3 hood, 20BT breathing tube, F30 flow control Tychem QC
19-321-721 RT4 hood, RTBT breathing tube, waist strap Tychem SL
17-989-623 RT4 hood, 20BT breathing tube, F30 flow control Tychem SL
17-989-621 RT1 hood, 20BT breathing tube, F30 flow control Tychem QC
17-989-622 RT2 hood, 20BT breathing tube, F30 flow control Tychem SL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-161-222 Supplier Bullard™ Supplier No. RT1DA
Add to Cart
Add to Cart
This item is not returnable. View return policy
Delivers compressed air through certified breathing air hose to workers wearing facemasks
RT Series Hood can be used for both Powered Air-Purifying Respirators (PAPR) and Supplied Air Respirators (SAR).
  • Headband-free design
  • Choice of Tychem QC or SL
  • 1000 Assigned Protection Factor (APF)
  • Panoramic viewing lens
  • Sport neck cuff
  • Long, double bibs
  • Taped and sealed seams (RT4 only)
  • No fit testing needed
The RT Series Hoods are available in several configurations to meet most levels of disposability requirements. As with most Tychem hoods, the RT Series is available as a stand-alone replacement hood for those applications that allow for decontamination and reuse of the breathing tube. For single use applications, the RT Series is available fully assembled with a disposable lightweight breathing tube. Individually packaged with the breathing tube already attached, the user needs only to don the hood and connect the flow control device. After exiting from the area, the user may dispose of the complete assembly.
Specifications
Certifications/Compliance FDA
Material Tychem QC
Product Type Supplied Air Respirator Assembly
For Use With (Equipment) Supplied air respirator
Includes RT3 hood, RTBT breathing tube
Model RT1DA
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.