Learn More
Bullard™ Tychem™ QC Replacement Hood, Threaded Connection
SDP
Catalog No. 19321725 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Tychem QC
Catalog No. Material
19-321-725 Tychem QC
1 options
Catalog No. 19-321-725 Supplier Bullard™ Supplier No. RT3T
Add to Cart
Add to Cart
Quick-attach T-Series hood. RT Style DuPont™ Tychem™ QC double-bib hood features patented air delivery system directs air to breathing zone and prevents fogging.
  • Inner Bib
  • 20 in. long outer bib
  • Headband-free design
  • Extra-large, panoramic viewing lens
  • No suspension required
  • Threaded tube connection; requires a different breathing tube SARHBT(supplied air) PAHBT (PAPR)
See also:  Model RT3 with sport neck cuff.
Specifications
Material Tychem QC
Product Type Respirator Hood
Model RT3T
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.