Learn More
Bullard™ Tychem™ SL Replacement Hood, Threaded Connection
SDP
Catalog No. 19321730 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Tychem SL
Catalog No. Material
19-321-730 Tychem SL
1 options
Catalog No. 19-321-730 Supplier Bullard™ Supplier No. RT4T
Add to Cart
Add to Cart
Double bib SAR and PAPR replacement hood features acetate lens, sport cuff, sealed seams, and threaded quick-attach connection.
  • Made of DuPont™ Tychem SL is Saranex-coated, features taped and sealed seams and provides excellent permeation characteristics against more caustic chemicals
  • 135 sq. in. lens provides for unobstructed visibility
  • Long inner and outer bibs for splash protection
  • Independent front and rear inner bibs help eliminate shoulder fit issues
Specifications
Connection Type Threaded
Color White
Material Tychem SL
Product Type Hood Assembly
Model RT4T
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.