Learn More
Bullard™ Six-Point Suspension Ratchet Caps
SDP
Catalog No. 19807755 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Hi-vis Orange
Navy Blue with AKZO Nobel Logo
Soft Green with AKZO Nobel Logo
White with AKZO Nobel Logo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-807-755 Hi-vis Orange
19-807-751 White with AKZO Nobel Logo
19-807-752 Soft Green with AKZO Nobel Logo
19-807-753 Navy Blue with AKZO Nobel Logo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-807-755 Supplier Bullard™ Supplier No. 30HOR
  • $37.60
    $25.65 / Each
    Save $11.95
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $653.50
    $445.65 / Case of 20 EA
    Save $207.85
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Classic cap style with comfortable six-point suspension.
  • Classic Series exceeds ANSI/ISEA Z89.1-2014, Type 1, Class E and G requirements, and meets CSA Z94.1-2005, Type I, Class E and G requirement
  • Available in different colors
Specifications
Certifications/Compliance Exceeds ANSI/ISEA Z89.1-2014, Type 1, Class E and G requirements, and meets CSA Z94.1-2005, Type I, Class E and G requirements
Color Hi-vis Orange
Style Classic
Suspension Type Six-point
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.