Learn More
Bullard™ Spectrum PD and PDE Coupler Assembly
SDP
Catalog No. 19824364 Shop All Bullard Company Products
Change view
Click to view available options
Material:
Brass
Schrader
Stainless Steel
Steel
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-824-364 Stainless Steel
19-824-366 Schrader
19-824-367 Steel
19-824-365 Stainless Steel
19-824-368 Brass
19-824-369 Stainless Steel
19-824-370 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-824-364 Supplier Bullard™ Supplier No. CA30
Specifications
Connection Type Industrial Interchange, Hansen
Material Stainless Steel
Product Type Coupler Assembly
For Use With (Equipment) Spectrum PD and PDE
Includes Male threaded nipple, female threaded coupler and adapter
Model CA30
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.