Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Sportek™ Accessories and Replacement Parts for Hard Hat
SDP

Catalog No. 19084043 Shop All Bullard Company Products
Change view
Click to view available options
Brow pad
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-084043 Brow pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-084043 Supplier Bullard™ Supplier No. TT3

missing translation for 'validMainframeMsg'

For use with face protection

Specifications

Color Blue
For Use With (Equipment) 4 pt. suspension hard hats
Product Type Brow pad

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.