Learn More
Bullard™ Standard Series S51 Caps
SDP
Catalog No. 19098029 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Black
Blue
Charcoal Brown
Forest Green
Gray
Hi-vis Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Navy Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Pin-Lock
4-Point Ratchet
Flex-Gear Ratchet
Standard
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-098-029 Hi-vis Yellow 4-Point Ratchet
19-070857 Tan Flex-Gear Ratchet
17-359-1D Blue Flex-Gear Ratchet
17-359-1B Yellow Flex-Gear Ratchet
17-359-1C Red Flex-Gear Ratchet
19-098-028 Hi-vis Orange 4-Point Ratchet
19-160-477 Charcoal Brown Flex-Gear Ratchet
19-160-479 Pacific Blue Flex-Gear Ratchet
19-160-481 Black Flex-Gear Ratchet
17-359-1A White Flex-Gear Ratchet
19-807-804 Forest Green Flex-Gear Ratchet
19-072427 Gray Flex-Gear Ratchet
19-098-024 Hi-vis Green 4-Point Pin-Lock
19-098-026 Hi-vis Yellow 4-Point Pin-Lock
19-098-027 Hi-vis Green 4-Point Ratchet
19-080663 Kelly Green 4-Point Pin-Lock
19-822-170 Navy Blue 4-Point Pin-Lock
17-359B Yellow Standard
17-359C Red Standard
17-359D Blue Standard
17-986-226 Orange Standard
19-021431 Forest Green Standard
19-098-025 Hi-vis Orange 4-Point Pin-Lock
19-160-475 Tan 4-Point Pin-Lock
19-160-474 Pacific Blue 4-Point Pin-Lock
19-160-476 Black 4-Point Pin-Lock
17-359A White Standard
19-807-811 White 4-Point Pin-Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Catalog No. 19-098-029 Supplier Bullard™ Supplier No. 51HYR
Add to Cart
Add to Cart
Dielectric hard cap is one of the lightest safety caps available
  • Constructed of polyethylene
  • Soft poly browpad absorbs moisture
  • Rain trough provides extra protection from snow, rain, dust and dirt
  • Accessory slots accommodate a variety of optional attachments
  • Available with standard four-point suspension or four-point Flex-Gear™ ratchet suspension featuring two vertical adjustments
  • Wide 0.75 in. (1.9cm) crown straps and low nape strap keep caps securely in place
Order Info
Note: There is a minimum order quantity of 20.
Compliance
ANSI/ISEA Z89.1-2009, Type I, Class E & G and CSA Z94.1-2005, Type 1, Class E & G requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C
Color Hi-vis Yellow
Suspension Type 4-Point Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.