Learn More
Bullard™ Standard Series S51 Caps
SDP
Supplier:  Bullard™ 51DGP
Catalog No. 19-160-473
Description
Description
  • Constructed of polyethylene
  • Soft poly browpad absorbs moisture
  • Rain trough provides extra protection from snow, rain, dust and dirt
  • Accessory slots accommodate a variety of optional attachments
  • Available with standard four-point suspension or four-point Flex-Gear™ ratchet suspension featuring two vertical adjustments
  • Wide 0.75 in. (1.9cm) crown straps and low nape strap keep caps securely in place
Product Suggestions
Product Suggestions