Learn More
Bullard™ Standard Series S61 Unvented Hardhats
SDP
Catalog No. 19160442 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Black
Charcoal Brown
Dove Gray
Forest Green
Hi-vis Orange
Hi-vis Yellow
Kelly Green
Kentucky Blue
Orange
Pacific Blue
Red
Tan
White
Yellow
Suspension Type:
4-Point Flex-Gear Ratchet
4-Point Pin-Lock
Flex-Gear Ratchet
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Suspension Type
19-160-442 Black Flex-Gear Ratchet
19-160-433 Kentucky Blue 4-Point Flex-Gear Ratchet
19-160-436 Hi-vis Orange 4-Point Flex-Gear Ratchet
19-160-438 Kelly Green Flex-Gear Ratchet
19-160-432 Forest Green Flex-Gear Ratchet
19-160-439 Dove Gray Flex-Gear Ratchet
19-160-435 Charcoal Brown Flex-Gear Ratchet
19-160-440 Pacific Blue Flex-Gear Ratchet
19-160-417 Orange 4-Point Pin-Lock
19-160-415 White 4-Point Pin-Lock
19-160-418 Forest Green 4-Point Pin-Lock
19-160-416 Red 4-Point Pin-Lock
19-160-420 Yellow 4-Point Pin-Lock
19-160-425 Dove Gray 4-Point Pin-Lock
19-160-422 Hi-vis Orange 4-Point Pin-Lock
19-160-423 Hi-vis Yellow 4-Point Pin-Lock
19-160-421 Charcoal Brown 4-Point Pin-Lock
19-160-427 Tan 4-Point Pin-Lock
19-160-426 Pacific Blue 4-Point Pin-Lock
19-160-428 Black 4-Point Pin-Lock
19-160-429 White 4-Point Flex-Gear Ratchet
19-160-441 Tan Flex-Gear Ratchet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
This item is not returnable. View return policy
Catalog No. 19-160-442 Supplier Bullard™ Supplier No. 61BKR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Low-profile shell design with four-point suspension system
Setting “The Standard”. Sleek, low-profile design with a patented “Flex-Gear” Ratchet suspension option.
Features
  • Absorbent, replaceable cotton brow pad is pillowed for maximum comfort
  • Ideal for construction and everyday work environments
  • One of the lightest helmets of its class
  • Unvented; see Model S62 for vented style
  • Accessory slots allow attachment of accessories such as earmuffs and face protection
  • Vertical height adjustment capability (2 front, 2 rear)
Pinlock or Ratchet Suspension
  • Choose 4-point pinlock suspension or 4-point “Flex-Gear” ratchet suspension
  • Pinlock suspension provides incremental sizing of 1/16 hat size (5mm), for a secure fit
  • Ratchet suspension can be adjusted between hat sizes 61/2-8
Order Info
Snap-on replacement brow pads are available (see below).
Compliance
ANSI Z89.1-2003, Type I, Class E and G (S61 Model); Class C (S62 Vented).
Specifications
Certifications/Compliance ANSI/ISEA Z89.1 Type I, Class E & G
Color Black
Suspension Type Flex-Gear Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.