Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ 88VX Series Golden Gate Style Nylon Cape
SDP

Catalog No. 189994433 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Tan
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-999-4433 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-4433 Supplier Bullard™ Supplier No. 21VX
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tan nylon replacement cape for use with 88VX supplied-air respirator systems.

Specifications

Color Tan
For Use With (Equipment) Supplied Air Respirator
Material Nylon
Includes Snap Hooks, Breathing Tube Strap, Additional Nylon Liner in Neck Cuff Area
Model 21VX
Overall Length (English) 38 in.
Overall Length (Metric) 96.5 cm
Product Type Cape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.