Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ Supplied-Air Respirator Accessories, Outer Lens
SDP

Catalog No. 19824421 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-824-421 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-824-421 Supplier Bullard™ Supplier No. MB1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PETG outer lens for use with GR50 supplied-air respirators.

Specifications

Color Blue
For Use With (Equipment) GR50 Airline Respirator
Model MB1
Product Type Outer Lens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.