Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bullard™ PAPR Tychem QC Dust Cover for Motor Blower
SDP

Catalog No. 19321652 Shop All Bullard Company Products
Change view
Click to view available options
Dust cover for PAPR and sleeve for breathing tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-321-652 Dust cover for PAPR and sleeve for breathing tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-321-652 Supplier Bullard™ Supplier No. PAC2QC

Dust cover for use with PA30/40 PAPR systems.

Specifications

Color White
Disposable Reusable
Material Tychem QC
Product Type Blower Cover
For Use With (Equipment) PA30 and 40 PAPR Systems
Model PAC2QC

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.