Learn More
Bullard™ PC90 Supplied-Air Respirator Breathing Tube
SDP
Catalog No. 19824383 Shop All Bullard Company Products
Change view
Click to view available options
Breathing tube for adjustable flow control valves
Catalog No.
19-824-383 Breathing tube for adjustable flow control valves
1 options
Catalog No. 19-824-383 Supplier Bullard™ Supplier No. 90BT
Breathing tube component for use with PC90 series respirators.
  • Component for use with PC 90 series respirators
  • Breathing tubes must be used in conjunction with hood, flow control device, and air hose
Specifications
Product Type Breathing Tube
For Use With (Equipment) RT Series supplied-air respirators
Includes S18051 hose clamp
Model 90BT
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.