Learn More
Bullard™ Tritan™ Chemical and Impact Resistant Visors
SDP
Catalog No. 19021471 Shop All Bullard Company Products
This item is not returnable. View return policy
Catalog No. 19-021-471 Supplier Bullard™ Supplier No. 840T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Impact protection and chemical resistance in one faceshield
Designed to help provide protection to the face from many hazards
Tough
  • Impact-resistant
  • Shatter-resistant
  • Reliable clarity
Durable
  • Chemical-resistant
  • Stain-resistant
  • High-temperature resistant
Sustainability
  • Unique chemistry
  • Bisphenol A (BPA) free
  • Estrogenic Activity (EA) free
When used with any Bullard bracket, all visors conform to ANSI/ISEA Z87.1-2010, Z87+ impact protector, standard for faceshields.
Refineries, laboratories, grinding
Compliance
Meets ANSI/ISEA Z87.1 and ANSI Z87 standards.
Specifications
Shape Heat-formed
Width (English) 15 in.
Lens Tint Clear
Length (English) 8 in.
No. per Case 50/Cs.