Learn More
Bullard™ Tychem™ Respirator Hoods with Hard Hats
SDP
Catalog No. 18998015 Shop All Bullard Company Products
Change view
Click to view available options
Tychem SL; W/C30R hat; 19 in. bib
Catalog No.
18-998-015 Tychem SL; W/C30R hat; 19 in. bib
1 options
This item is not returnable. View return policy
Catalog No. 18-998-015 Supplier Bullard™ Supplier No. 20SICVHC30R
This item is not returnable. View return policy
Specifications
Assigned Protection Factor (APF) 1,000
Material Tychem SL
Product Type Headpiece
For Use With (Equipment) Pa30 series powered air purifying respirator
Includes Double bib hood, taped seams, extra long, C30R hard hat
Model 20SICVHC30R
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.