Learn More
Bullard™ V10 Series Starter Hose Kits
SDP
Catalog No. 18998077 Shop All Bullard Company Products
Change view
Click to view available options
Color:
Black
Green
Length (English):
25 ft.
50 ft.
Connection Type:
Schrader, Steel quick-disconnect Coupler
Snap-Tite, Interchange Coupler
Snap-Tite, Steel quick-disconnect Coupler
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Connection Type
18-998-077 Green 50 ft. Snap-Tite, Steel quick-disconnect Coupler
18-998-075 Black 25 ft. Snap-Tite, Steel quick-disconnect Coupler
18-998-076 Green 25 ft. Snap-Tite, Interchange Coupler
18-998-078 Black 25 ft. Schrader, Steel quick-disconnect Coupler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-998-077 Supplier Bullard™ Supplier No. 46917
Add to Cart
Add to Cart
Kit includes one V10 starter hose with female quick-disconnect coupler on one end and standard female hose thread on the other.
  • Connect the respirator’s breathing tube fitting to the female quick disconnect coupler
  • choose from differet hose lengths and quick disconnect coupler options
  • Extension hose kit includes:
    • One V10 extension hose with female fittings on both ends
    • One V11 3/8” hose-to-3/8” hose adaptor
    • One V13 3/8” hose-to-3/8” pipe adaptor
Specifications
Connection Type Snap-Tite, Steel quick-disconnect Coupler
Color Green
Product Type Hose Kit
Includes V10 starter hose, female quick-disconnect coupler, standard female hose thread, V13 hose-to-pipe adaptor
Model 46917
Length (English) 50 ft.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.