Learn More
Cayman Chemical Bullatine B, Purity: ≥98%, CAS: 466-26-2, 10 mg
SDP
Catalog No. 502911780
Encompass_Preferred
A diterpene alkaloid; inhibits Nav1.7 in HEK293 cells expressing the human channel (IC50 = 25.8 nM); scavenges DPPH radicals by 65.3% at 1 mM; increases the mechanical pain threshold in the von Frey test in streptozotocin-induced diabetic, but not non-diabetic, mice at 10 mg/kg
Catalog No. 50-291-1780 Supplier Cayman Chemical Supplier No. 3617010 MG
A diterpene alkaloid; inhibits Nav1.7 in HEK293 cells expressing the human channel (IC50 = 25.8 nM); scavenges DPPH radicals by 65.3% at 1 mM; increases the mechanical pain threshold in the von Frey test in streptozotocin-induced diabetic, but not non-diabetic, mice at 10 mg/kg