Learn More
Roboz Surgical Roboz Bulldog Clamp, Curved, 1.5 Inch, 500-900g Pressure, 1 Ea
SDP
Catalog No. 5000574728
Encompass
This curved surgical bulldog clamp is designed for precise vessel occlusion in research applications. Measuring 35 mm (1.5 in) in length, it delivers consistent pressure ranging from 500 to 900 grams to ensure secure closure during experimental procedures. The instrument features a non-riveted design for specialized surgical utility.
  • Curved jaw profile for optimal access
  • Provides 500 to 900 grams of occlusive pressure
  • Constructed without a rivet
  • Designed specifically for research and laboratory use
Catalog No. 50-005-74728 Supplier Roboz Surgical Supplier No. RS744135
This curved surgical bulldog clamp is designed for precise vessel occlusion in research applications. Measuring 35 mm (1.5 in) in length, it delivers consistent pressure ranging from 500 to 900 grams to ensure secure closure during experimental procedures. The instrument features a non-riveted design for specialized surgical utility.
  • Curved jaw profile for optimal access
  • Provides 500 to 900 grams of occlusive pressure
  • Constructed without a rivet
  • Designed specifically for research and laboratory use