Learn More
Roboz Surgical Roboz Johns Hopkins Bulldog Clamp, Curved, 2 in, 1 ea
SDP
Catalog No. 5000574764
Encompass
This Johns Hopkins bulldog clamp is a surgical instrument designed for temporary vessel occlusion during delicate procedures. Featuring a curved geometry and a length of 50 mm, this device provides precise control and stable compression. Please note this product is not for human use.
  • Curved shape for improved access in surgical sites
  • Total length of 50 mm (2 in.)
  • Pressure rating of 800 g
  • Constructed without a rivet
  • Designed for laboratory or research applications only
Catalog No. 50-005-74764 Supplier Roboz Surgical Supplier No. RS744350
This Johns Hopkins bulldog clamp is a surgical instrument designed for temporary vessel occlusion during delicate procedures. Featuring a curved geometry and a length of 50 mm, this device provides precise control and stable compression. Please note this product is not for human use.
  • Curved shape for improved access in surgical sites
  • Total length of 50 mm (2 in.)
  • Pressure rating of 800 g
  • Constructed without a rivet
  • Designed for laboratory or research applications only