Learn More
World Precision Instrument Dieffenbach Bulldog Clamp, 6.5cm, Straight
SDP
Catalog No. 50822224
Encompass
Dieffenbach Bulldog Clamps are a versatile surgical instrument used for precise and reliable occlusion (blocking) of blood vessels or tissue to control bleeding during surgical procedures. These clamps are designed for pinching delicate or small blood vessel. Dieffenback Bulldog Clamps have a spring-loaded mechanism and a series of sharp teeth on the clamp's jaws. The teeth help to grip the blood vessel or tissue firmly, while the spring-loaded mechanism keeps the clamp closed. These clamps are commonly used in vascular surgeries, including open-heart surgery, where they are used to clamp off the coronary artery during a bypass procedure.
Catalog No. 50-822-224 Supplier World Precision Instrument Supplier No. 14116
Dieffenbach Bulldog Clamps are a versatile surgical instrument used for precise and reliable occlusion (blocking) of blood vessels or tissue to control bleeding during surgical procedures. These clamps are designed for pinching delicate or small blood vessel. Dieffenback Bulldog Clamps have a spring-loaded mechanism and a series of sharp teeth on the clamp's jaws. The teeth help to grip the blood vessel or tissue firmly, while the spring-loaded mechanism keeps the clamp closed. These clamps are commonly used in vascular surgeries, including open-heart surgery, where they are used to clamp off the coronary artery during a bypass procedure.