Learn More
World Precision Instrument Micro Bulldog Clamp, 16cm, Serrated, Curved Jaw, German
SDP
Catalog No. 50822231
Encompass
Micro Bulldog Clamps are surgical instruments used in microsurgery and delicate surgical procedures to occlude small blood vessels or tissue to control bleeding. Micro bulldog clamps are smaller in size compared to regular Diefenbach Bulldog clamps, which makes them well-suited for use in microsurgery and other delicate procedures. The teeth on the clamp's jaws are designed to be atraumatic, meaning they are less sharp than those found on larger Diefenbach Bulldog clamps. This helps to minimize tissue damage during occlusion of small blood vessels. Our bulldog clamps come with either straight or curved jaws. They have a spring-loaded mechanism that keeps the jaws closed, occluding the blood vessel or tissue. You pinch them together to open the jaws and position them. When you let go, they stay closed, clamping the tissue. Micro Bulldog Clamps are reusable and should be thoroughly cleaned and sterilized promptly after each use.
Catalog No. 50-822-231 Supplier World Precision Instrument Supplier No. 14119G
Micro Bulldog Clamps are surgical instruments used in microsurgery and delicate surgical procedures to occlude small blood vessels or tissue to control bleeding. Micro bulldog clamps are smaller in size compared to regular Diefenbach Bulldog clamps, which makes them well-suited for use in microsurgery and other delicate procedures. The teeth on the clamp's jaws are designed to be atraumatic, meaning they are less sharp than those found on larger Diefenbach Bulldog clamps. This helps to minimize tissue damage during occlusion of small blood vessels. Our bulldog clamps come with either straight or curved jaws. They have a spring-loaded mechanism that keeps the jaws closed, occluding the blood vessel or tissue. You pinch them together to open the jaws and position them. When you let go, they stay closed, clamping the tissue. Micro Bulldog Clamps are reusable and should be thoroughly cleaned and sterilized promptly after each use.