Learn More
Fine Science Tools BULLDOG SERREFINE-CURVED 50MM
SDP
Catalog No. NC0118187
Encompass
Product: Bulldog Serrefine - Curved/50mm/ Shape: Curved/ Jaw Dimensions: 15.75 x 2.3mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 50mm/ Serrated: Yes/ Unit: Each
Catalog No. NC0118187 Supplier Fine Science Tools Supplier No. 1805150
Product: Bulldog Serrefine - Curved/50mm/ Shape: Curved/ Jaw Dimensions: 15.75 x 2.3mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 50mm/ Serrated: Yes/ Unit: Each