Description
- Includes 1 sturdy black kit case:
- 10 Step 1 lead solvent ampoules
- 10 Step 2 lead reagent ampoules
- 10 Step 3 copper solvent ampoules
- 10 Step 4 copper reagent ampoules
- 50 Benchkote test papers
- 100 plastic bags for packing positive test papers
- 4 technician report forms
- 1 white photographic scale
- 1 permanent marker
- Helps determine the caliber and direction in which a bullet was fired
- Simple Four-step process that takes 3-4 minutes from sampling to final results
- Allows several bullet holes to be analyzed using one set of test tubes
- Materials Includes:
- Plastic-backed filter paper
- Glass ampoules
- Plastic tubes
- Options and Accessories
- FS472: Benchkote Testing Paper Refill
- FS473: Step 1 Lead Solvent Ampoule Refill
- FS474: Step 2 Lead Reagent Ampoule Refill
- FS475: Step 3 Copper Solvent Ampoule Refill
- FS476: Step 4 Copper Reagent Ampoule Refill
Specifications
Specifications
| Product Type | Master Kit |
| Dimensions (L x W x H) | 13.1 x 10.4 x 3.33 in. |