Learn More
Lonza Walkersville SkGM-2 Skeletal Muscle Cell Growth Medium-2 BulletKit
Catalog No. NC9993177
Encompass
We offer two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 BulletKit (CC-3245) is an enhancement of the SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM BulletKit (CC-3160) is optimized for use with human skeletal muscle cells. SkGM -2is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x SkBM-2 Basal Medium (CC-3246), 500 mL1 x SkGM-2 SingleQuots Supplement Pack (CC-3244) containing:1 x Red Cap Vial with GA-1000, 0.50 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Natural Cap Vial with Dexamethasone, 0.50 mL1 x Bottle FBS, 50.00 mL1 x Bottle L-Glutamine, 10.00 mL
Catalog No. NC9993177 Supplier Lonza Walkersville Supplier No. CC3245
We offer two varieties of Skeletal Muscle Growth Medium - SkGM and SkGM-2.SkGM-2 Skeletal Muscle Cell Growth Medium-2 BulletKit (CC-3245) is an enhancement of the SkGM Medium. Fetuin has been removed and there has been further optimization of the basal media.SkGM BulletKit (CC-3160) is optimized for use with human skeletal muscle cells. SkGM -2is specially designed for use with normal and diabetic skeletal muscle myoblasts.Content:1 x SkBM-2 Basal Medium (CC-3246), 500 mL1 x SkGM-2 SingleQuots Supplement Pack (CC-3244) containing:1 x Red Cap Vial with GA-1000, 0.50 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Natural Cap Vial with Dexamethasone, 0.50 mL1 x Bottle FBS, 50.00 mL1 x Bottle L-Glutamine, 10.00 mL