Learn More
Lonza Walkersville EGM Endothelial Cell Growth Medium BulletKit
Supplier:  Lonza Walkersville CC3124
Encompass
Choose from a variety of endothelial cell growth media for an easy cultivation of primary endothelial cells. Every endothelial cell growth medium has its special formulation and is optimized to grow certain cell types. Each type of endothelial cell growth medium is guaranteed to perform when used to grow Clonetics Primary Cells. Content:1 x EBM Basal Medium (CC-3121), 500 mL1 x EGM SingleQuots Supplement Pack (CC-4133) containing:1 x Green Cap Vial with hEGF 0.50 mL1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle BBE 2.00 mL1 x Blue Cap Vial with Ascorbic Acid 0.50 mL1 x Bottle FBS 10.00 mL
Catalog No. NC9291749