Learn More
Selleck Chemical LLC Bulleyaconi cine A S3907-25mg
SDP
Catalog No. 503108313
Encompass_Preferred
Bulleyaconi cine A (Bulleyaconitine A BLA) is an active ingredient of Aconitum bulleyanum plants and classified as an aconitine-like alkaloid It is a potent use-dependent blocker for both Nav1 7 and Nav1 8 Na currents
Catalog No. 50-310-8313 Supplier Selleck Chemical LLC Supplier No. S390725MG
Bulleyaconi cine A (Bulleyaconitine A BLA) is an active ingredient of Aconitum bulleyanum plants and classified as an aconitine-like alkaloid It is a potent use-dependent blocker for both Nav1 7 and Nav1 8 Na currents