Learn More
Midwest Scientific Bullseye Agarose General Purpose - standard electrophoresis, 500g
SDP
Catalog No. NC0800285
Encompass
Bulls Eye Agarose GP2 is highly refined and highly purified. Each lot is physically tested and verified for performance in its designated application. General purpose electrophoresis and blotting. Excellent separation, resolution, and clarity. Low absorption and background from staining reagents. High gel strength for safe and easy handling. Highly refined and purified. Greater thermal stability. Packaged with spec sheet, prep instructions, and MSDS. 500g
Catalog No. NC0800285 Supplier Midwest Scientific Supplier No. BEA500
Bulls Eye Agarose GP2 is highly refined and highly purified. Each lot is physically tested and verified for performance in its designated application. General purpose electrophoresis and blotting. Excellent separation, resolution, and clarity. Low absorption and background from staining reagents. High gel strength for safe and easy handling. Highly refined and purified. Greater thermal stability. Packaged with spec sheet, prep instructions, and MSDS. 500g