Learn More
Midwest Scientific PR1MA SmartGlow DNA Safe Stain, 1mL1 Pre-stain, Blue and UV Light
SDP
Catalog No. NC1550783
Encompass
PR1MA SmartStain PS (Pre-Stain) can be used as a direct replacement for Ethidium Bromide in agarose and polyacrylamide gel electrophoresis. The stain emits green fluorescence when bound to dsDNA or ssDNA and emits red fluorescence when bound to RNA. PR1MA SmartStain PS exhibits excitation peaks at 290 nm and 490 nm, allowing it to be used with UV and blue light.
Catalog No. NC1550783 Supplier Midwest Scientific Supplier No. BE4500PS
PR1MA SmartStain PS (Pre-Stain) can be used as a direct replacement for Ethidium Bromide in agarose and polyacrylamide gel electrophoresis. The stain emits green fluorescence when bound to dsDNA or ssDNA and emits red fluorescence when bound to RNA. PR1MA SmartStain PS exhibits excitation peaks at 290 nm and 490 nm, allowing it to be used with UV and blue light.