Learn More
Electron Microscopy Sciences Tough-Spots Bullseye Label, 7/16", Dark Green
SDP
Catalog No. 5034519
Encompass_Preferred
Tough-tags make labeling and identifying samples quick and easy are temperature resistant from -196 degree C to 80 degree C. Each ToughSpot has a different color to identify similar samples and also includes a white area for a note.
Size: 7/16"
Catalog No. 50-345-19 Supplier Electron Microscopy Sciences Supplier No. 7759311
Tough-tags make labeling and identifying samples quick and easy are temperature resistant from -196 degree C to 80 degree C. Each ToughSpot has a different color to identify similar samples and also includes a white area for a note.
Size: 7/16"