Learn More
Medchemexpress LLC Buloxibutid (AT2 receptor agonist C21) | 477775-14-7 | MFCD26936332 | 96.8% | 475.62 | C23H29N3O4S2 | 50 MG
SDP
Catalog No. 5000215780
Encompass_Preferred
Buloxibutid is a selective angiotensin II type 2 (AT2) receptor agonist (C21) used in research to study AT2-mediated GPCR signaling and related biological effects. It exhibits high affinity for AT2 and minimal activity at AT1, with manufacturer-provided data for solubility and storage to support experimental use.
  • Selective AT2 receptor agonist with Ki 0.4 nM
  • Markedly lower affinity for AT1 receptor (>10 μM)
  • Soluble in DMSO at 50 mg/mL for in vitro applications
  • In vivo formulation guidance provided with multiple solvent sequences tested
  • Powder stable at -20°C for long-term storage; solution storage limits specified
Catalog No. 50-002-15780 Supplier Medchemexpress LLC Supplier No. HY10011350MG
Buloxibutid is a selective angiotensin II type 2 (AT2) receptor agonist (C21) used in research to study AT2-mediated GPCR signaling and related biological effects. It exhibits high affinity for AT2 and minimal activity at AT1, with manufacturer-provided data for solubility and storage to support experimental use.
  • Selective AT2 receptor agonist with Ki 0.4 nM
  • Markedly lower affinity for AT1 receptor (>10 μM)
  • Soluble in DMSO at 50 mg/mL for in vitro applications
  • In vivo formulation guidance provided with multiple solvent sequences tested
  • Powder stable at -20°C for long-term storage; solution storage limits specified