Learn More
MSC Bulwark Coverall, Lightweight Excel FR ComforTouch, 9.1 ATPV, 1 ea
Catalog No. 5000687728
Encompass
This lightweight uniform coverall provides CAT 2 FR/AR protection without added bulk. It includes innovative features for extra comfort and convenience. All pockets are designed to keep personal effects safe.
  • One-piece, topstitched, lay-flat collar
  • Two-way, concealed Nomex® taped brass break-away zipper
  • Concealed snap at top of zipper at neck
  • Concealed snap on cuff
  • Elastic waist inserts
  • Deep-pleated action back
  • Side vent openings with snap closure
Catalog No. 50-006-87728 Supplier MSC Supplier No. 66652868
This lightweight uniform coverall provides CAT 2 FR/AR protection without added bulk. It includes innovative features for extra comfort and convenience. All pockets are designed to keep personal effects safe.
  • One-piece, topstitched, lay-flat collar
  • Two-way, concealed Nomex® taped brass break-away zipper
  • Concealed snap at top of zipper at neck
  • Concealed snap on cuff
  • Elastic waist inserts
  • Deep-pleated action back
  • Side vent openings with snap closure