Learn More
MSC Bulwark Coverall, Midweight, Excel FR, CAT 2 Protection, 1 ea
Catalog No. 5000769739
Encompass
This midweight classic coverall provides CAT 2 FR/AR protection, designed for comfort and utility in demanding work environments. It features five well-placed pockets for tool storage and side vents for breathability.
  • One-piece, topstitched, lay-flat collar
  • Two-way concealed Nomex® taped brass break-away zipper with concealed snap at neck
  • Hemmed sleeves
  • Side vent openings
  • Two set-in front pockets, two patch hip pockets, one chest pocket
  • Arc Rating ATPV 11 cal/cm², Category 2 Protection
  • NFPA® 2112 Compliant
  • Flame resistant, 9 oz. twill fabric (100% cotton)
Catalog No. 50-007-69739 Supplier MSC Supplier No. 39822606
This midweight classic coverall provides CAT 2 FR/AR protection, designed for comfort and utility in demanding work environments. It features five well-placed pockets for tool storage and side vents for breathability.
  • One-piece, topstitched, lay-flat collar
  • Two-way concealed Nomex® taped brass break-away zipper with concealed snap at neck
  • Hemmed sleeves
  • Side vent openings
  • Two set-in front pockets, two patch hip pockets, one chest pocket
  • Arc Rating ATPV 11 cal/cm², Category 2 Protection
  • NFPA® 2112 Compliant
  • Flame resistant, 9 oz. twill fabric (100% cotton)