Learn More
Medchemexpress LLC Bumetanide | 28395-03-1 | 99.9% | 364.42 | 50 MG
SDP
Catalog No. 5000386607
Encompass_Preferred
Bumetanide (Standard) is the analytical standard of Bumetanide. It is a highly potent loop diuretic and a Na+-K+-Cl+ cotransporter (NKCC) blocker, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Highly potent loop diuretic
  • Na+-K+-Cl+ cotransporter (NKCC) blocker
  • Selective NKCC1 inhibitor
  • Inhibits NKCC2
  • Intended for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-86607 Supplier Medchemexpress LLC Supplier No. HY17468R50MG
Bumetanide (Standard) is the analytical standard of Bumetanide. It is a highly potent loop diuretic and a Na+-K+-Cl+ cotransporter (NKCC) blocker, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Highly potent loop diuretic
  • Na+-K+-Cl+ cotransporter (NKCC) blocker
  • Selective NKCC1 inhibitor
  • Inhibits NKCC2
  • Intended for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS