Learn More
Medchemexpress LLC HY-17468 500mg Medchemexpress, Bumetanide CAS:28395-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17468500MG
Encompass_Preferred
Medchemexpress, HY-17468 500mg Bumetanide CAS:28395-03-1 Bumetanide (Ro 10-6338;PF 1593) is a selective Na+-K+-Cl- cotransporter 1 (NKCC1) inhibitor, weakly inhibits NKCC2, with IC50s of 0.68 and 4.0 μM for hNKCC1A and hNKCC2A, respectively[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0777