Learn More
Enzo Life Sciences Bumetanide (5g). CAS: 28395-03-1
SDP
Supplier:  Enzo Life Sciences BMLEI1680005
Encompass_Preferred
Bumetanide is a specific inhibitor of Na-K-2Cl cotransport. It binds specifically, saturably and with high affinity to membranes from a large variety of cells. A clinically useful diuretic. Purity: ≥98%. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0584