Learn More
Apexbio Technology LLC Bumetanide, 5g Cas# 28395-03-1
SDP
Supplier:  Apexbio Technology LLC A18555000
Encompass_Preferred
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter. More potent than furosemide. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8513