Learn More
Medchemexpress LLC Bumetanide-d5 | 1216739-35-3 | 99.8% | 369.45 | 5 MG
SDP
Catalog No. 5000386706
Encompass_Preferred
Bumetanide-d5 is a deuterium-labeled Bumetanide. It functions as a selective Na+-K+-Cl- (NKCC1) inhibitor and weakly inhibits NKCC2, with IC50s of 0.68 and 4.0 μM for hNKCC1A and hNKCC2A, respectively. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Bumetanide
  • Selective Na+-K+-Cl- (NKCC1) inhibitor
  • Weakly inhibits NKCC2
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-86706 Supplier Medchemexpress LLC Supplier No. HY17468S5MG
Bumetanide-d5 is a deuterium-labeled Bumetanide. It functions as a selective Na+-K+-Cl- (NKCC1) inhibitor and weakly inhibits NKCC2, with IC50s of 0.68 and 4.0 μM for hNKCC1A and hNKCC2A, respectively. This compound is suitable for use as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Bumetanide
  • Selective Na+-K+-Cl- (NKCC1) inhibitor
  • Weakly inhibits NKCC2
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Appearance: Solid
  • Color: White to off-white