Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Bump Cap

Catalog No. p-7230454
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-185-009 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-185-009 Supplier Honeywell™ Supplier No. HBCE080000

Enhanced protection against impact risks and superior comfort both at work and after; excellent quality, high performance and great design when hard hats are not required

  • Water-proof coating and hand-washable (cotton/polyester)
  • Reflective material for increased visibility
  • 2.16 in. peak brim for better visibility
  • Ultra-soft foam for extra comfort
  • Polypropylene protective shell for ultimate protection
  • Velcro™ adjustment for multiple head sizes
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Application) Manufacturing services, Food, Utilities, Agriculture, Transportation, Weekend warriors and to-doers
Material Cotton/Polyester
Product Type Bump Cap
Size Universal
Ventilated Ventilated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.