Learn More
Grainger Bumper 1-1/2in. Black Rubber PK10
Catalog No. 502766231
Encompass_Preferred
The 16VC06 is a 16-ounce clear plastic squeeze bottle with a black continuous thread closure made by Berry Plastics. It has a 6-400 neck finish and is made from PETE plastic. This bottle has a capacity of 16 fluid ounces, stands 6.5 inches tall, has a 2.5 inch diameter, and features graduations up to 16 ounces. The closure has a tamper-evident ring and the bottles are sold in cases of 24. This versatile bottle can be used for a variety of liquids including soaps, lotions, cleaners, and more.
Catalog No. 50-276-6231 Supplier Grainger Supplier No. 16VC06
The 16VC06 is a 16-ounce clear plastic squeeze bottle with a black continuous thread closure made by Berry Plastics. It has a 6-400 neck finish and is made from PETE plastic. This bottle has a capacity of 16 fluid ounces, stands 6.5 inches tall, has a 2.5 inch diameter, and features graduations up to 16 ounces. The closure has a tamper-evident ring and the bottles are sold in cases of 24. This versatile bottle can be used for a variety of liquids including soaps, lotions, cleaners, and more.