Learn More
Grainger Bumper 1-1/2in. Black Rubber PK10
Catalog No. 502766232
Encompass_Preferred
The Grainger 16VC07 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. This product is VOC compliant and made in the USA.
Catalog No. 50-276-6232 Supplier Grainger Supplier No. 16VC07
The Grainger 16VC07 is a 16-ounce aerosol can of CRC Brakleen non-chlorinated brake parts cleaner. It is a fast drying, non-flammable cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. This product is VOC compliant and made in the USA.