Learn More
Grainger Bumper 1-1/2in. Black Rubber PK10
Catalog No. 502766386
Encompass_Preferred
The Grainger 16VC05 is a Dayton 1/3 HP Centrifugal Pump with a 56Y frame, 1725 RPM, 115V, 60Hz, 1 phase. It has a thermoplastic body and impeller, and a stainless steel shaft. The pump has a max flow of 30 GPM, max head of 13 feet, and max pressure of 5.7 PSI. It has a 1-1/4" NPT inlet and 1" NPT outlet. The pump weighs 9 lbs and has dimensions of 5-3/4" length x 4-1/2" width x 4-3/4" height. It can handle temperatures up to 180°F.
Catalog No. 50-276-6386 Supplier Grainger Supplier No. 16VC05
The Grainger 16VC05 is a Dayton 1/3 HP Centrifugal Pump with a 56Y frame, 1725 RPM, 115V, 60Hz, 1 phase. It has a thermoplastic body and impeller, and a stainless steel shaft. The pump has a max flow of 30 GPM, max head of 13 feet, and max pressure of 5.7 PSI. It has a 1-1/4" NPT inlet and 1" NPT outlet. The pump weighs 9 lbs and has dimensions of 5-3/4" length x 4-1/2" width x 4-3/4" height. It can handle temperatures up to 180°F.