Learn More
Grainger Bumper 1/2 in Black Rubber PK25
Catalog No. 502764224
Encompass_Preferred
The Grainger Approved Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a hole diameter of 1", an overall height of 2-1/2", and an overall width of 2-1/2". This black bumper is made of SBR rubber and has a durometer of 60A. It can withstand temperatures between -20 to 180 degrees Fahrenheit. The bumper has a tensile strength of 2000 psi and meets military specification MIL-P-81528. It comes in a convenient pack of 4 bumpers. This high quality bumper absorbs shock and reduces noise in industrial equipment and machinery.
Catalog No. 50-276-4224 Supplier Grainger Supplier No. 16WC74
The Grainger Approved Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a hole diameter of 1", an overall height of 2-1/2", and an overall width of 2-1/2". This black bumper is made of SBR rubber and has a durometer of 60A. It can withstand temperatures between -20 to 180 degrees Fahrenheit. The bumper has a tensile strength of 2000 psi and meets military specification MIL-P-81528. It comes in a convenient pack of 4 bumpers. This high quality bumper absorbs shock and reduces noise in industrial equipment and machinery.