Learn More
Grainger Bumper 1/2 in Black Rubber PK25
Catalog No. 502764406
Encompass_Preferred
The Grainger 16WD08 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/2" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door with two included screws. It has a black finish and is designed to protect walls and door surfaces by absorbing the impact of a swinging door. This product is made in the USA and is commonly used in commercial buildings.
Catalog No. 50-276-4406 Supplier Grainger Supplier No. 16WD08
The Grainger 16WD08 Bumper Door Stop is a door-mounted door stop made of rubber and steel. It has a diameter of 1-1/2", projects 2-1/2" from the door surface, and withstands up to 100 lbs of holding force. This door stop features a mounting bracket that attaches to the door with two included screws. It has a black finish and is designed to protect walls and door surfaces by absorbing the impact of a swinging door. This product is made in the USA and is commonly used in commercial buildings.