Learn More
Grainger Bumper 1/2 in Black Rubber PK25
Catalog No. 502767884
Encompass_Preferred
The 1/2" Black Rubber Bumper from Grainger is a rubber bumper with a 1/2" diameter and 1/2" height. It has a black finish and is made from SBR rubber. The bumper has a durometer hardness of 60A. It can be used as a rubber foot to reduce vibration, a spacer, or stop. The operating temperature range is -20°F to 180°F. This rubber bumper is sold individually.
Catalog No. 50-276-7884 Supplier Grainger Supplier No. 16ZD31
The 1/2" Black Rubber Bumper from Grainger is a rubber bumper with a 1/2" diameter and 1/2" height. It has a black finish and is made from SBR rubber. The bumper has a durometer hardness of 60A. It can be used as a rubber foot to reduce vibration, a spacer, or stop. The operating temperature range is -20°F to 180°F. This rubber bumper is sold individually.