Learn More
Grainger Bumper 1/2 in Black Rubber PK25
Catalog No. 502768046
Encompass_Preferred
The Grainger Approved Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a 2" diameter, is 3" long, has a 1/4" mounting hole, and is made of SBR rubber. This black bumper has applications including material handling equipment, conveyors, and dock equipment. It meets ASTM D2000 M4BA710 A13 B13 F17 standards. The operating temperature range is -20°F to 180°F.
Catalog No. 50-276-8046 Supplier Grainger Supplier No. 16ZD58
The Grainger Approved Bumper is a rubber bumper that provides impact absorption and noise reduction. It has a 2" diameter, is 3" long, has a 1/4" mounting hole, and is made of SBR rubber. This black bumper has applications including material handling equipment, conveyors, and dock equipment. It meets ASTM D2000 M4BA710 A13 B13 F17 standards. The operating temperature range is -20°F to 180°F.