Learn More
Grainger Bumper 1/2 in Black Rubber PK50
Catalog No. 502763672
Encompass_Preferred
The 16WD04 is a 16-inch, 4-blade aluminum propeller by Sensenich Propeller Manufacturing Company, designed for aircraft engines up to 200 horsepower. Made from 6061-T6 forged aluminum alloy, it ensures strength and durability. Precision CNC machining provides tight tolerances and smooth operation. Approved under FAA-PMA regulations, its large diameter and low pitch offer excellent low-end torque and climb performance. The corrosion-resistant aluminum blades ensure long service life, making the 16WD04 ideal for many light aircraft applications.
Catalog No. 50-276-3672 Supplier Grainger Supplier No. 16WD04
The 16WD04 is a 16-inch, 4-blade aluminum propeller by Sensenich Propeller Manufacturing Company, designed for aircraft engines up to 200 horsepower. Made from 6061-T6 forged aluminum alloy, it ensures strength and durability. Precision CNC machining provides tight tolerances and smooth operation. Approved under FAA-PMA regulations, its large diameter and low pitch offer excellent low-end torque and climb performance. The corrosion-resistant aluminum blades ensure long service life, making the 16WD04 ideal for many light aircraft applications.