Learn More
Grainger Bumper 1/2 in Black Rubber PK50
Catalog No. 502764724
Encompass_Preferred
The Grainger Approved 16WC42 Bumper is a black rubber bumper with a 2-1/2" diameter and a 1-1/4" width. It has a 1/4" diameter center hole and is made of SBR rubber. This bumper absorbs shock and protects surfaces from damage. It can be used on carts, racks, conveyors and other material handling equipment to prevent metal-to-metal contact. The operating temperature range is -20°F to 180°F.
Catalog No. 50-276-4724 Supplier Grainger Supplier No. 16WC42
The Grainger Approved 16WC42 Bumper is a black rubber bumper with a 2-1/2" diameter and a 1-1/4" width. It has a 1/4" diameter center hole and is made of SBR rubber. This bumper absorbs shock and protects surfaces from damage. It can be used on carts, racks, conveyors and other material handling equipment to prevent metal-to-metal contact. The operating temperature range is -20°F to 180°F.